Learn More
OHAUS 2 Foam Inserts
Catalog No. 01804335 Shop All OHAUS Products
Change view
Click to view available options
Holds:
14 to 29 mm Tubes
29 to 37 mm Tubes
9 to 16 mm Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
01-804-335 9 to 16 mm Tubes
01-804-383 14 to 29 mm Tubes
01-804-405 29 to 37 mm Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-804-335 Supplier OHAUS Supplier No. 83041431
Add to Cart
Add to Cart
Two high-density Foam Inserts to accommodate 14-29 mm, 29-37 mm, 9-16 mm diameter tubes.
  • The insert fits into the Recessed Platform 504-0039-00, for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
TRUSTED_SUSTAINABILITY
Specifications
Holds 9 to 16 mm Tubes
Product Type Vortex Attachment
Model Recessed Platform