Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 2-Formyl-5-methoxyphenylboronic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-chloro-2-butanone reacts with 1,4-dianion of acetophenone n-ethoxycarbonylhydrazone to yield pyrazoline derivatives.3-chloro-2-butanone was used in production of chiral alcohols from acetophenone derivatives, α-ketoesters and n-boc-3-pyrrolidinone by recombinant e
- Coli cells
- It was used in the synthesis of carbene precursor, 3-aryl-4,5-dimethylthiazolium chloride
Specifications
Specifications
| CAS | 40138-18-9 |
| Molecular Formula | C8H9BO4 |
| Linear Formula | C8H9BO4 |
| MDL Number | MFCD03001335 |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 179.97 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?