Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Formylbenzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AAH2884603
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2884603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2884603 Supplier Thermo Scientific Chemicals Supplier No. H2884603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 380151-85-9
Density 1.053
Molecular Formula C13H17BO3
Refractive Index 1.513
Quantity 1g
MDL Number MFCD07363841
Sensitivity Air Sensitive
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-formylphenylboronic acid pinacol ester, 2-formylphenyl boronic acid pinacol ester, 2-formylphenylboronic acid, pinacol ester, 2-formylbenzeneboronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-2-formylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzaldehyde, amtb063, 2-boronobenzaldehyde, pinacol ester
InChI Key SLJMPQLPRHIAOM-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C=O
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular Weight (g/mol) 232.086
PubChem CID 10977322
Formula Weight 232.08
Percent Purity 97%
Chemical Name or Material 2-Formylbenzeneboronic acid pinacol ester
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.