Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B18735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1873-5G 5 g
B18731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1873-5G Supplier TCI America Supplier No. B18735G
Add to Cart
Add to Cart

Specifications

CAS 40138-16-7
Melting Point 125°C
Molecular Formula C7H7BO3
Quantity 5 g
MDL Number MFCD00151822
Synonym 2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde
InChI Key DGUWACLYDSWXRZ-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C=O)(O)O
IUPAC Name (2-formylphenyl)boronic acid
Molecular Weight (g/mol) 149.94
PubChem CID 292189
Formula Weight 149.94
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.