Learn More
Thomas Scientific 2" White Freezer Box w/ 100 Place Divider, No Drain Holes, 5 1/4" x 5 1/4" x 2"
SDP
Catalog No. NC0253466
Encompass
Without Drain Holes, Premium Frost Resistant Box, Suitable for -80°C or Vapor Phase LN2
Catalog No. NC0253466 Supplier Thomas Scientific Supplier No. 1149H27
Without Drain Holes, Premium Frost Resistant Box, Suitable for -80°C or Vapor Phase LN2