Learn More
Medchemexpress LLC 2-Furancarboxamide, 3-(3,4-difluoro-2-methoxyphenyl)-N-[6-[(1R)-1,2-dihydroxyethyl]-3-pyridin... | 2877755-11-6 | 99.9% | 490.42 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY17124410MM/1ML
Encompass_Preferred
Sodium Channel inhibitor 6 (compound I) is a selective Nav1.8 inhibitor that can be used in the study of neuropathic pain. It is an intermediate for synthesizing potent Nav1.8 inhibitors.
  • Selective Nav1.8 inhibitor
  • Used in the study of neuropathic pain
  • Intermediate for synthesizing potent Nav1.8 inhibitors
Catalog No. 50-003-86506