Learn More
Medchemexpress LLC 2-Furancarboxamide, 3-(3,4-difluoro-2-methoxyphenyl)-N-[6-[(1R)-1,2-dihydroxyethyl]-3-pyridinyl... | 2877755-11-6 | 929055 | 99.9% | 490.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1712441MG
Encompass_Preferred
Sodium Channel inhibitor 6 (compound I) is a selective Nav1.8 inhibitor that can be used in the study of neuropathic pain. It is an intermediate for synthesizing potent Nav1.8 inhibitors. This product is for research use only.
  • Selective Nav1.8 inhibitor.
  • Used in the study of neuropathic pain.
  • Intermediate for synthesizing potent Nav1.8 inhibitors.
  • High purity: 99.93%.
Catalog No. 50-003-86454