Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Furancarboxamide, 5-cyano-N-(2,5-di-1-piperidinylphenyl)- | 959626-45-0 | 99.6% | 378.47 | 1 ML
SDP

Catalog No. 5000438259
Encompass_Preferred

cFMS Receptor Inhibitor IV is a chemical compound with the molecular formula C22H26N4O2 and a molecular weight of 378.47. It appears as a light yellow to yellow solid and has a purity of 99.61%. This inhibitor is consistent with its structure as confirmed by 1H NMR Spectrum and LCMS analysis.

  • High purity of 99.61% (LCMS)
  • Consistent with structure (1H NMR Spectrum and LCMS)
  • Appearance: light yellow to yellow solid
  • Storage conditions: 4°C (protect from light); in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)

Catalog No. 50-004-38259 Supplier Medchemexpress LLC Supplier No. HY11245510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cFMS Receptor Inhibitor IV is a chemical compound with the molecular formula C22H26N4O2 and a molecular weight of 378.47. It appears as a light yellow to yellow solid and has a purity of 99.61%. This inhibitor is consistent with its structure as confirmed by 1H NMR Spectrum and LCMS analysis.

  • High purity of 99.61% (LCMS)
  • Consistent with structure (1H NMR Spectrum and LCMS)
  • Appearance: light yellow to yellow solid
  • Storage conditions: 4°C (protect from light); in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.