Learn More
Medchemexpress LLC 2-Furancarboxamide, 5-nitro-N-[4-[7-(1,2,3,6-tetrahydro-4-pyridinyl)imidazo[1,2-a]pyridin-3-yl]phenyl] | 25 MG
SDP
Catalog No. 5000375340
Encompass_Preferred
W1131 TFA is a potent STAT3 inhibitor that induces ferroptosis. It inhibits cancer progression in subcutaneous xenograft, organoid, and PDX models of gastric cancer. It effectively alleviates cancer cell chemoresistance to 5-FU, and regulates the cell cycle, DNA damage response, oxidative phosphorylation, the IL6-JAK-STAT3 pathway, and the ferroptosis pathway.
  • Potent STAT3 inhibitor
  • Induces ferroptosis
  • Inhibits cancer progression in subcutaneous xenograft, organoid, and PDX models of gastric cancer
  • Alleviates cancer cell chemoresistance to 5-FU
  • Regulates the cell cycle
  • Regulates DNA damage response
  • Regulates oxidative phosphorylation
  • Regulates the IL6-JAK-STAT3 pathway
  • Regulates the ferroptosis pathway
Catalog No. 50-003-75340 Supplier Medchemexpress LLC Supplier No. 25MGHY153190A
W1131 TFA is a potent STAT3 inhibitor that induces ferroptosis. It inhibits cancer progression in subcutaneous xenograft, organoid, and PDX models of gastric cancer. It effectively alleviates cancer cell chemoresistance to 5-FU, and regulates the cell cycle, DNA damage response, oxidative phosphorylation, the IL6-JAK-STAT3 pathway, and the ferroptosis pathway.
  • Potent STAT3 inhibitor
  • Induces ferroptosis
  • Inhibits cancer progression in subcutaneous xenograft, organoid, and PDX models of gastric cancer
  • Alleviates cancer cell chemoresistance to 5-FU
  • Regulates the cell cycle
  • Regulates DNA damage response
  • Regulates oxidative phosphorylation
  • Regulates the IL6-JAK-STAT3 pathway
  • Regulates the ferroptosis pathway