Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Furfurylthio)ethylamine 98.0+%, TCI America™
SDP

Catalog No. F03785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03785G 5 g
F037825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03785G Supplier TCI America Supplier No. F03785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36415-21-1
Molecular Formula C7H11NOS
Molecular Weight (g/mol) 157.231
MDL Number MFCD01075029
InChI Key VSPDYEHKAMKDNW-UHFFFAOYSA-N
Synonym (2-Aminoethylthio)methylfuran
PubChem CID 118970
IUPAC Name 2-(furan-2-ylmethylsulfanyl)ethanamine
SMILES C1=COC(=C1)CSCCN

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 157.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Furfurylthio)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.