Learn More
Medchemexpress LLC 2-Furoic acid | 88-14-2 | 99.98% | 112.08 | 1 G
SDP
Catalog No. 5000454083
Encompass_Preferred
2-Furoic acid, also known as Furan-2-carboxylic acid, is an organic compound that is produced through the oxidation of furfural. It has been observed to have hypolipidemic effects, reducing both serum cholesterol and serum triglyceride levels in rats.
  • It is an organic compound produced via furfural oxidation.
  • It exhibits hypolipidemic effects.
  • It lowers both serum cholesterol and serum triglyceride levels in rats.
  • It significantly reduces liver and small intestine ATP dependent citrate lyase, acetyl CoA synthetase, acyl CoA cholesterol acyl transferase, sn-glycerol 3-phosphate acyl transferase, phosphatidylate phosphohydrolase, and heparin induced lipoprotein lipase activities.
  • The product appears as a white to off-white solid.
Catalog No. 50-004-54083 Supplier Medchemexpress LLC Supplier No. HYW0129461G
2-Furoic acid, also known as Furan-2-carboxylic acid, is an organic compound that is produced through the oxidation of furfural. It has been observed to have hypolipidemic effects, reducing both serum cholesterol and serum triglyceride levels in rats.
  • It is an organic compound produced via furfural oxidation.
  • It exhibits hypolipidemic effects.
  • It lowers both serum cholesterol and serum triglyceride levels in rats.
  • It significantly reduces liver and small intestine ATP dependent citrate lyase, acetyl CoA synthetase, acyl CoA cholesterol acyl transferase, sn-glycerol 3-phosphate acyl transferase, phosphatidylate phosphohydrolase, and heparin induced lipoprotein lipase activities.
  • The product appears as a white to off-white solid.