Learn More
Medchemexpress LLC 2-Furoic acid-d3 | 40073-83-4 | 99.99% | 115.10 | 25 MG
SDP
Catalog No. 5000452046
Encompass_Preferred
2-Furoic acid-d3 is the deuterium labeled 2-Furoic acid. 2-Furoic acid (Furan-2-carboxylic acid) is an organic compound produced through furfural oxidation. 2-Furoic acid exhibits hypolipidemic effect, lowering both serum cholesterol and serum triglyceride levels in rats.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52046 Supplier Medchemexpress LLC Supplier No. 25MGHYW012946S
2-Furoic acid-d3 is the deuterium labeled 2-Furoic acid. 2-Furoic acid (Furan-2-carboxylic acid) is an organic compound produced through furfural oxidation. 2-Furoic acid exhibits hypolipidemic effect, lowering both serum cholesterol and serum triglyceride levels in rats.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.