Learn More
CPC Scientific 2-Furoyl-Leu-Ile-Gly-Arg-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110090
Encompass
PAR2 Tethered Ligand (Murine) / Protease-Activated Receptor 2 (PAR2) AgonistONE-LETTER SEQUENCE: 2-Furoyl-LIGRL-NH2MOLECULAR FORMULA: C31H52N9O7MOLECULAR WEIGHT:662.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Hematology
Catalog No. 50-211-0090 Supplier CPC Scientific Supplier No. PAP002B
PAR2 Tethered Ligand (Murine) / Protease-Activated Receptor 2 (PAR2) AgonistONE-LETTER SEQUENCE: 2-Furoyl-LIGRL-NH2MOLECULAR FORMULA: C31H52N9O7MOLECULAR WEIGHT:662.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Hematology