Learn More
CPC Scientific 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110087
Encompass
Potent and Selective Protease-Activated Receptor 2 (PAR2) AgonistSEQUENCE: 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: 2-Furoyl-LIGRL-Orn-NH2MOLECULAR FORMULA: C36H62N11O8MOLECULAR WEIGHT: 777.97STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [729589-58-6]SYNONYMS: (2-Furoyl)-PAR-2 (2-6)-Orn amide, (2-Furoyl)-PAR-2 (2-6)-Orn amide (mouse, rat), 2-Furoyl-LIGRLO-NH2 Potent and Selective Protease-Activated Receptor 2 (PAR2) Agonist, (2-Furoyl)-LIGRLOamideRESEARCH AREA: HematologyREFERENCES:SKU(s): PAP-001A, PAP-001B
Catalog No. 50-211-0087 Supplier CPC Scientific Supplier No. PAP001A
Potent and Selective Protease-Activated Receptor 2 (PAR2) AgonistSEQUENCE: 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: 2-Furoyl-LIGRL-Orn-NH2MOLECULAR FORMULA: C36H62N11O8MOLECULAR WEIGHT: 777.97STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [729589-58-6]SYNONYMS: (2-Furoyl)-PAR-2 (2-6)-Orn amide, (2-Furoyl)-PAR-2 (2-6)-Orn amide (mouse, rat), 2-Furoyl-LIGRLO-NH2 Potent and Selective Protease-Activated Receptor 2 (PAR2) Agonist, (2-Furoyl)-LIGRLOamideRESEARCH AREA: HematologyREFERENCES:SKU(s): PAP-001A, PAP-001B