Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furoylacetonitrile, 97%

Catalog No. AAA1449714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1449714 25 g
AAA1449703 1 g
AAA1449706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1449714 Supplier Thermo Scientific Chemicals Supplier No. A1449714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 31909-58-7 | C7H5NO2 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31909-58-7
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00052210
InChI Key RZNSHBXVTAHWPP-UHFFFAOYSA-N
Synonym 2-furoylacetonitrile, 3-furan-2-yl-3-oxopropanenitrile, 3-2-furyl-3-oxopropanenitrile, 2-cyanoacetylfuran, 2-furanoylacetonitrile, 2-furoyl acetonitrile, pubchem11959, acmc-1cock, 2-2-furanoyl acetonitrile
PubChem CID 141671
IUPAC Name 3-(furan-2-yl)-3-oxopropanenitrile
SMILES O=C(CC#N)C1=CC=CO1

Specifications

Melting Point 77°C to 80°C
Quantity 25 g
Beilstein 111902
Formula Weight 135.12
Percent Purity 97%
Chemical Name or Material 2-Furoylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.