Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F03945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0394-5G 5 g
F03941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0394-5G Supplier TCI America Supplier No. F03945G
Add to Cart
Add to Cart

Specifications

CAS 13331-23-2
Molecular Formula C4H5BO3
MDL Number MFCD00799544
Quantity 5 g
Synonym 2-furanboronic acid, furan-2-boronic acid, 2-furylboronic acid, 2-furanylboronic acid, furan-2-ylboranediol
InChI Key PZJSZBJLOWMDRG-UHFFFAOYSA-N
SMILES B(C1=CC=CO1)(O)O
IUPAC Name furan-2-ylboronic acid
Molecular Weight (g/mol) 111.891
PubChem CID 2734357
Formula Weight 111.89
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.