Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furylmethyl isothiocyanate, ≥95%, Thermo Scientific™

Catalog No. SB01625EB
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SB01625EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01625EB Supplier Thermo Scientific Chemicals Supplier No. SB01625EB

Specifications

CAS 4650-60-6
CAS Min % 95.0
Density 1.3900g/mL
Assay Percent Range 95%+
Molecular Formula C6H5NOS
Quantity 25g
MDL Number MFCD00014112
Synonym furfuryl isothiocyanate, 2-isothiocyanatomethyl furan, 2-furylmethyl isothiocyanate, 2-isothiocyanatomethyl-furan, furan, 2-isothiocyanatomethyl, 2-furanylmethyl isothiocyanate, 2-furylmethanisothiocyanate, furfurylisothiocyanate, furylmethylisothiocyanate, 7-bromo-quinolin-2-ylamine
InChI Key ICXYINJACKJQQV-UHFFFAOYSA-N
SMILES C1=COC(=C1)CN=C=S
IUPAC Name 2-(isothiocyanatomethyl)furan
Molecular Weight (g/mol) 139.172
PubChem CID 78380
Formula Weight 139.18
Percent Purity 95%
Chemical Name or Material 2-Furylmethyl isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.