Learn More
Medchemexpress LLC HY-103667 5mg Medchemexpress, 2-HBA CAS:131359-24-5 Purity:>98%
SDP
Catalog No. 501968157
Encompass_Preferred
Medchemexpress, HY-103667 5mg 2-HBA CAS:131359-24-5 2-HBA is a potent inducer of NAD(P)H:quinone acceptor oxidoreductase 1 (NQO1) which can also activate caspase-3 and caspase-10. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8157 Supplier Medchemexpress LLC Supplier No. HY1036675MG
Medchemexpress, HY-103667 5mg 2-HBA CAS:131359-24-5 2-HBA is a potent inducer of NAD(P)H:quinone acceptor oxidoreductase 1 (NQO1) which can also activate caspase-3 and caspase-10. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.