Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptadecylimidazole 85.0+%, TCI America™
SDP

Catalog No. H0021500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0021500G 500 g
H002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0021500G Supplier TCI America Supplier No. H0021500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23328-87-2
Molecular Formula C20H38N2
Molecular Weight (g/mol) 306.538
MDL Number MFCD00038620
InChI Key YTWBFUCJVWKCCK-UHFFFAOYSA-N
PubChem CID 90068
IUPAC Name 2-heptadecyl-1H-imidazole
SMILES CCCCCCCCCCCCCCCCCC1=NC=CN1

Specifications

Melting Point 89°C
Quantity 500 g
Formula Weight 306.54
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Heptadecylimidazole
EINECSNumber (5)-0383
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.