Learn More
Medchemexpress LLC 2-heptanol | 543-49-7 | MFCD00004587 | 99.7% | 116.2 g/mol | C7H16O | 1 KG
SDP
Catalog No. 5000299920
Encompass_Preferred
2-Heptanol (heptan-2-ol) is a secondary alcohol used as a research reagent and analytical reference. It occurs naturally in essential oils and is applied in antimicrobial, metabolic, and flavor chemistry studies. Supplied at high analytical purity for laboratory and analytical applications.
  • High analytical purity suitable for gas chromatography and analytical work.
  • Useful in antimicrobial and metabolic research applications.
  • Compatible with common organic solvents, including DMSO.
  • Molecular formula C7H16O; molar mass 116.2 g/mol.
  • Available in bulk quantities for laboratory-scale use.
Catalog No. 50-002-99920 Supplier Medchemexpress LLC Supplier No. HYW0158791KG
2-Heptanol (heptan-2-ol) is a secondary alcohol used as a research reagent and analytical reference. It occurs naturally in essential oils and is applied in antimicrobial, metabolic, and flavor chemistry studies. Supplied at high analytical purity for laboratory and analytical applications.
  • High analytical purity suitable for gas chromatography and analytical work.
  • Useful in antimicrobial and metabolic research applications.
  • Compatible with common organic solvents, including DMSO.
  • Molecular formula C7H16O; molar mass 116.2 g/mol.
  • Available in bulk quantities for laboratory-scale use.