Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-heptanol | 543-49-7 | MFCD00004587 | 99.7% | 116.20 | C7H16O | 25 G
SDP

Catalog No. 5000303576
Encompass_Preferred

2-Heptanol (CAS 543-49-7) is a secondary alcohol used as a chemical reagent, analytical standard, and solvent in research and development. It is a clear, colorless liquid with documented antimicrobial activity in some studies and is commonly used in analytical chemistry, flavor and fragrance applications, and biochemical assays.

  • High purity: 99.65% (manufacturer reported)
  • Molecular formula C7H16O and molecular weight 116.20
  • Available in multiple pack sizes, including 25 g
  • Soluble in common organic solvents and DMSO (solutions available)
  • Suitable as reagent, analytical standard, and reference material

Catalog No. 50-003-03576 Supplier Medchemexpress LLC Supplier No. 25GHYW015879
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Heptanol (CAS 543-49-7) is a secondary alcohol used as a chemical reagent, analytical standard, and solvent in research and development. It is a clear, colorless liquid with documented antimicrobial activity in some studies and is commonly used in analytical chemistry, flavor and fragrance applications, and biochemical assays.

  • High purity: 99.65% (manufacturer reported)
  • Molecular formula C7H16O and molecular weight 116.20
  • Available in multiple pack sizes, including 25 g
  • Soluble in common organic solvents and DMSO (solutions available)
  • Suitable as reagent, analytical standard, and reference material

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.