Learn More
Medchemexpress LLC 2-heptanol | 543-49-7 | MFCD00004587 | 99.7% | 116.20 g·mol⁻¹ | C7H16O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01587910MM/1ML
Encompass_Preferred
2-Heptanol is a secondary alcohol supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research and analytical applications. It serves as a reagent or reference standard in biological assays and analytical workflows, offering high purity and a convenient liquid format for accurate dosing and rapid use.
Catalog No. 50-002-99595