Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-heptanol | 543-49-7 | MFCD00004587 | 99.7% | 116.20 g/mol | C7H16O | 100 G
SDP

Catalog No. 5000303763
Encompass_Preferred

2-Heptanol is a secondary alcohol (heptan-2-ol) used as a reagent and analytical standard in research applications. It is a colorless to light-yellow liquid (CAS 543-49-7) commonly used in essential-oil analysis, antimicrobial studies, and chromatographic methods.

  • High purity: 99.65%.
  • Physical form: colorless to light-yellow liquid, density 0.817 g/cm3.
  • Suitable as an analytical standard and reagent for research use.
  • Offered in multiple pack sizes, including 100 g, for laboratory needs.
  • Soluble in common organic solvents; available in DMSO solution formulations.

Catalog No. 50-003-03763 Supplier Medchemexpress LLC Supplier No. HYW015879100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Heptanol is a secondary alcohol (heptan-2-ol) used as a reagent and analytical standard in research applications. It is a colorless to light-yellow liquid (CAS 543-49-7) commonly used in essential-oil analysis, antimicrobial studies, and chromatographic methods.

  • High purity: 99.65%.
  • Physical form: colorless to light-yellow liquid, density 0.817 g/cm3.
  • Suitable as an analytical standard and reagent for research use.
  • Offered in multiple pack sizes, including 100 g, for laboratory needs.
  • Soluble in common organic solvents; available in DMSO solution formulations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.