Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptanol, 99+%

Catalog No. AC120372500
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120372500 250 mL Glass bottle
AC120370500 50 mL Glass bottle
AC120370010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC120372500 Supplier Thermo Scientific Chemicals Supplier No. 120372500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 543-49-7 | C7H16O | 116.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-49-7
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004587
InChI Key CETWDUZRCINIHU-UHFFFAOYNA-N
Synonym 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
PubChem CID 10976
IUPAC Name heptan-2-ol
SMILES CCCCCC(C)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.8175g/mL
Boiling Point 160°C to 162°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.422
Linear Formula CH3(CH2)4CH(OH)CH3
Quantity 250 mL
Beilstein 01, 415
Specific Gravity 0.8175
Merck Index 15, 4697
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,benzene,miscible with most common organic liquids
Formula Weight 116.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Heptanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or
EINECSNumber 208-844-3
RTECSNumber MJ2975000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.