Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Heptanone 98 fCC fG20KG
Catalog No. 502914151
Encompass
2-Heptanone is an aroma compound. It is a bioactive agent and has potential as a pesticide. Kinetics of release of 2-heptanone incorporated into lipid films or composite solid lipid particle (SLP) films have been investigated. 2-Heptanone is secreted by the mandibular glands of honeybees when they bite. It is reported to act as an anaesthetic in small arthropods such as wax moth larva (WML) and Varroa mites which are paralysed after a honeybee bite.
Catalog No. 50-291-4151 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W25440120KGK
2-Heptanone is an aroma compound. It is a bioactive agent and has potential as a pesticide. Kinetics of release of 2-heptanone incorporated into lipid films or composite solid lipid particle (SLP) films have been investigated. 2-Heptanone is secreted by the mandibular glands of honeybees when they bite. It is reported to act as an anaesthetic in small arthropods such as wax moth larva (WML) and Varroa mites which are paralysed after a honeybee bite.