Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptanone, 99%

Catalog No. p-7080585
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10200AE 100 mL
AAA10200AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA10200AE Supplier Thermo Scientific Chemicals Supplier No. A10200AE
Only null left
Add to Cart

CAS: 110-43-0 | C7H14O | 114.19 g/mol

2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(C)=O

Specifications

Melting Point -35°C
Density 0.82
Boiling Point 149°C to 150°C
Flash Point 47°C (116°F)
Odor Unpleasant
Refractive Index 1.409
Quantity 100 mL
UN Number UN1110
Beilstein 1699063
Merck Index 14,4663
Solubility Information Slightly miscible with water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 2-Heptanone
Show More Show Less
Hazard Category H226-H302+H332-H336
Hazard Statement GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-AMYL METHYL KETONE
EINECSNumber 203-767-1
RTECSNumber MJ5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.