Learn More
Medchemexpress LLC 2-Heptenoic acid 250mg
SDP
Catalog No. 5000204523
Encompass_Preferred
2-Heptenoic acid is an unsaturated fatty acid It is a clear colorless liquid with a strong odour and is commonly used in the flavor and fragrance industry for its pungent anise-like odor In addition it can be used as an intermediate in the synthesis of other organic compounds including pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial applications including food and beverage production cosmetics and personal care products
Catalog No. 50-002-04523 Supplier Medchemexpress LLC Supplier No. 250MGHYW107869
2-Heptenoic acid is an unsaturated fatty acid It is a clear colorless liquid with a strong odour and is commonly used in the flavor and fragrance industry for its pungent anise-like odor In addition it can be used as an intermediate in the synthesis of other organic compounds including pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial applications including food and beverage production cosmetics and personal care products