Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptenoic Acid (contains 3-Heptenoic Acid) 90.0+%, TCI America™
SDP

Catalog No. H04265ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04265ML Supplier TCI America Supplier No. H04265ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18999-28-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00014019
InChI Key YURNCBVQZBJDAJ-AATRIKPKSA-N
PubChem CID 5282709
ChEBI CHEBI:38364
IUPAC Name (E)-hept-2-enoic acid
SMILES CCCCC=CC(=O)O

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 mL
UN Number 3265
Formula Weight 128.17
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptenoic Acid (contains 3-Heptenoic Acid)
EINECSNumber (2)-0609
RTECSNumber MJ9650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.