Learn More
AdipoGen 2-Heptyl-3-hydroxyl-4-quinolon
SDP
Catalog No. 502560766
Encompass_Preferred
Chemical. CAS 108985-27-9. Formula C16H21NO2. MW 259.34. Synthetic. 2-heptyl-3-hydroxy-4-quinolone can function as an intercellular signal. Quorum sensing-regulated virulence factor used to induce and study the regulation of virulence genes such as those involved in iron scavenging. Quorum sensing is a signaling system used by bacteria to coordinate activity based upon their population density. The system involves the exchange of signaling molecules among bacteria via cell receptors.
Catalog No. 50-256-0766 Supplier AdipoGen Supplier No. CDXH0077M250
Chemical. CAS 108985-27-9. Formula C16H21NO2. MW 259.34. Synthetic. 2-heptyl-3-hydroxy-4-quinolone can function as an intercellular signal. Quorum sensing-regulated virulence factor used to induce and study the regulation of virulence genes such as those involved in iron scavenging. Quorum sensing is a signaling system used by bacteria to coordinate activity based upon their population density. The system involves the exchange of signaling molecules among bacteria via cell receptors.