Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptylcyclopentanone 99.0+%, TCI America™
SDP

Catalog No. H056925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H056925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H056925ML Supplier TCI America Supplier No. H056925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 137-03-1
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.31
MDL Number MFCD00019315
InChI Key PJXHBTZLHITWFX-UHFFFAOYNA-N
Synonym 2-heptylcyclopentanone, 2-n-heptylcyclopentanone, cyclopentanone, 2-heptyl, alismone, cyclopentanone, 2-n-heptyl, alpha-heptyl cyclopentanone, alpha-heptylcyclopentanone, 2-n-heptyl cyclopentanone, heptylcyclopentan-1-one, heptylcyclopentanone
PubChem CID 8710
IUPAC Name 2-heptylcyclopentan-1-one
SMILES CCCCCCCC1CCCC1=O

Specifications

Color Colorless
Boiling Point 264°C
Quantity 25 mL
Formula Weight 182.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptylcyclopentanone
RTECSNumber GY4950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.