Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptynoic Acid 97.0+%, TCI America™
SDP

Catalog No. H06641ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06641ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06641ML Supplier TCI America Supplier No. H06641ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1483-67-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00014372
InChI Key UEERQXQKEJPYBR-UHFFFAOYSA-N
PubChem CID 73869
IUPAC Name hept-2-ynoic acid
SMILES CCCCC#CC(=O)O

Specifications

Boiling Point 141°C
Quantity 1 mL
UN Number 3265
Formula Weight 126.16
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Heptynoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.