Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Hexamethyleneimino)ethanol 95%, Thermo Scientific™

Catalog No. AC135490250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC135490250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC135490250 Supplier Thermo Scientific Chemicals Supplier No. 135490250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20603-00-3
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
InChI Key VMRYMOMQCYSPHS-UHFFFAOYSA-N
Synonym 2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine
PubChem CID 88615
IUPAC Name 2-(azepan-1-yl)ethanol
SMILES C1CCCN(CC1)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.059g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Refractive Index 1.4830 to 1.4860 (20°C, 589nm)
Quantity 25 mL
Formula Weight 143.23
Percent Purity ≥94%
Physical Form Viscous Liquid
Chemical Name or Material 2-(Hexamethyleneimino)ethanol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.