Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexen-1-ylsuccinic Anhydride (cis- and trans- mixture) 96.0+%, TCI America™
SDP

Catalog No. H061625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H061625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H061625G Supplier TCI America Supplier No. H061625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10500-34-2
Molecular Formula C10H14O3
MDL Number MFCD00047131

Specifications

Color Yellow
Quantity 25 g
Formula Weight 182.22
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Hexen-1-ylsuccinic Anhydride (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.