Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexene, Mixture of cis and trans 97%, Thermo Scientific™

Catalog No. AC294810100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC294810100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294810100 Supplier Thermo Scientific Chemicals Supplier No. 294810100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-43-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z
PubChem CID 639661
IUPAC Name (E)-hex-2-ene
SMILES CCCC=CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.678
Infrared Spectrum Authentic
Refractive Index 1.3940 to 1.3980 (20°C, 589nm)
Linear Formula CH3CH2CH2CH=CHCH3
Quantity 10 mL
Beilstein 01, I, 89
Formula Weight 84.15
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-Hexene, mixture of cis and trans, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.