Learn More
eMolecules​ 86308-26-1 | 2-Hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | Ambeed | MFCD08457665 | 212.14 | C12H25BO2 | 98 | CCCCCCB1OC(C)(C)C(C)(C)O1 | 25g | 589946779
Catalog No. NC3813038 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | Ambeed | 86308-26-1 | MFCD08457665 | 212.140 | C12H25BO2 | 98.000 | CCCCCCB1OC(C)(C)C(C)(C)O1 | 25g | 589946779
Catalog No. NC3813038 Supplier eMolecules​ Supplier No. 589946779
2-Hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | Ambeed | 86308-26-1 | MFCD08457665 | 212.140 | C12H25BO2 | 98.000 | CCCCCCB1OC(C)(C)C(C)(C)O1 | 25g | 589946779