Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hexyl-4-pentynoic acid | 96017-59-3 | MFCD00946863 | 98.0% | 182.26 g·mol-1 | C11H18O2 | 25 MG
SDP

Catalog No. 5000229003
Encompass_Preferred

2-Hexyl-4-pentynoic acid is a small-molecule HDAC inhibitor and alkyne-bearing fatty acid used in epigenetics, neuroprotection, and bioorthogonal click-chemistry studies.

  • Contains terminal alkyne functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Used as a histone deacetylase inhibitor in research applications.
  • Purity ≥98.0% as reported by the manufacturer.
  • Molecular weight 182.26 g·mol-1; formula C11H18O2.
  • Provided in milligram-scale packs suitable for laboratory discovery and assay work.

Catalog No. 50-002-29003 Supplier Medchemexpress LLC Supplier No. HY11878325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hexyl-4-pentynoic acid is a small-molecule HDAC inhibitor and alkyne-bearing fatty acid used in epigenetics, neuroprotection, and bioorthogonal click-chemistry studies.

  • Contains terminal alkyne functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Used as a histone deacetylase inhibitor in research applications.
  • Purity ≥98.0% as reported by the manufacturer.
  • Molecular weight 182.26 g·mol-1; formula C11H18O2.
  • Provided in milligram-scale packs suitable for laboratory discovery and assay work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.