Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexyn-1-ol 97.0+%, TCI America™
SDP

Catalog No. H04655ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04655ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04655ML Supplier TCI America Supplier No. H04655ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 764-60-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00039540
InChI Key BTDWSZJDLLLTMI-UHFFFAOYSA-N
Synonym 2-hexyn-1-ol, pubchem13064, acmc-209p3i, ksc380a2d, 2-hexyn-1-ol,
PubChem CID 69818
IUPAC Name hex-2-yn-1-ol
SMILES CCCC#CCO

Specifications

Color Yellow
Boiling Point 167°C
Quantity 5 mL
UN Number 1987
Formula Weight 98.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexyn-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.