Learn More
Cole-Parmer Two-Hole Black Rubber Stoppers, Standard Size 6; 22/pk
Supplier:  Cole-Parmer 6299177
Encompass_Preferred
These two-hole black stoppers made with solid natural rubber stay pliable over a long period of time. Provide good tightness and adhesion, withstand laboratory conditions without hardening, and are ideal for use with nonaggressive or low concentration solutions.
Catalog No. NC1574732