Learn More
Cole-Parmer Cole-Parmer Two-Hole Black Rubber Stoppers, Standard Size 00; 113/Pk
Catalog No. NC1574724 Shop All Cole-Parmer Products
Encompass_Preferred
These two-hole black stoppers made with solid natural rubber stay pliable over a long period of time. Provide good tightness and adhesion, withstand laboratory conditions without hardening, and are ideal for use with nonaggressive or low concentration solutions. Each stopper has its size marked in raised figures. Low sulfur content. Withstand temperatures from –13 to 158°F (–25 to 70°C).
Catalog No. NC1574724 Supplier Cole-Parmer Supplier No. 6299170
These two-hole black stoppers made with solid natural rubber stay pliable over a long period of time. Provide good tightness and adhesion, withstand laboratory conditions without hardening, and are ideal for use with nonaggressive or low concentration solutions. Each stopper has its size marked in raised figures. Low sulfur content. Withstand temperatures from –13 to 158°F (–25 to 70°C).