Learn More
Chemglass Life Sciences Rubber Stopper, #13-1/2, Two Holes, Black
SDP
Catalog No. NC1328131
Encompass_Preferred
Stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250¿F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application.
Catalog No. NC1328131 Supplier Chemglass Life Sciences Supplier No. CG302823
Stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250¿F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application.