Learn More
Medchemexpress LLC 2-Hydrazinobenzothiazole | 615-21-4 | 99.95% | C7H7N3S | 25 G
SDP
Catalog No. 5000358913
Encompass_Preferred
2-Hydrazinobenzothiazole is a potent IDO1 inhibitor with an IC50 of 8 μM. It interacts with the IDO1 haem through hydrazine and has potential for immune system related diseases research. It appears as a white to off-white solid.
  • Potent IDO1 inhibitor with an IC50 of 8 μM
  • Interacts with IDO1 haem through hydrazine
  • Potential for immune system related diseases research
  • Appearance: solid
  • Color: white to off-white
  • For research use only
Catalog No. 50-003-58913 Supplier Medchemexpress LLC Supplier No. 25GHY78699
Add to Cart
Add to Cart
2-Hydrazinobenzothiazole is a potent IDO1 inhibitor with an IC50 of 8 μM. It interacts with the IDO1 haem through hydrazine and has potential for immune system related diseases research. It appears as a white to off-white solid.
  • Potent IDO1 inhibitor with an IC50 of 8 μM
  • Interacts with IDO1 haem through hydrazine
  • Potential for immune system related diseases research
  • Appearance: solid
  • Color: white to off-white
  • For research use only