Learn More
Medchemexpress LLC 2-Hydrazinobenzothiazole | 615-21-4 | 99.95% | C7H7N3S | 50 G
SDP
Catalog No. 5000434886
Encompass_Preferred
This product is a potent inhibitor of IDO1, with an IC50 of 8 μM. It functions by interacting with the enzyme's haem through its hydrazine component. This compound shows potential for research related to immune system diseases.
  • Potent inhibitory activity against a specific enzyme.
  • Interacts with the enzyme's active site via a hydrazine group.
  • Suitable for investigation into immune system-related conditions.
Catalog No. 50-004-34886 Supplier Medchemexpress LLC Supplier No. HY7869950G
This product is a potent inhibitor of IDO1, with an IC50 of 8 μM. It functions by interacting with the enzyme's haem through its hydrazine component. This compound shows potential for research related to immune system diseases.
  • Potent inhibitory activity against a specific enzyme.
  • Interacts with the enzyme's active site via a hydrazine group.
  • Suitable for investigation into immune system-related conditions.