Learn More
Medchemexpress LLC 2-hydroxy-1-naphthaldehyde | 708-06-5 | MFCD00004005 | 99.9% | 172.18 g/mol | C11H8O2 | 100 G
SDP
Catalog No. 5000300430
Encompass_Preferred
CFL-137 (2-hydroxy-1-naphthaldehyde) is a small-molecule inhibitor selective for the KRasG12C mutant. It displays antiproliferative and anticancer activity in preclinical studies and is used as a tool compound to investigate KRas-driven signaling and lung cancer models. Characterized by CAS 708-06-5 and a molecular weight of 172.18 g/mol, it is supplied at high purity for research use.
  • Potent KRasG12C inhibition demonstrated in cell assays.
  • Shows antiproliferative activity and anticancer effects in preclinical models.
  • Suitable as a research tool for KRas-driven signaling and lung cancer studies.
  • High purity (99.92%), minimizing impurities in experimental work.
  • Well-characterized physical properties (C11H8O2, 172.18 g/mol).
  • Available in gram-scale quantities for synthesis and biological testing.
Catalog No. 50-003-00430 Supplier Medchemexpress LLC Supplier No. HYW016889100G
CFL-137 (2-hydroxy-1-naphthaldehyde) is a small-molecule inhibitor selective for the KRasG12C mutant. It displays antiproliferative and anticancer activity in preclinical studies and is used as a tool compound to investigate KRas-driven signaling and lung cancer models. Characterized by CAS 708-06-5 and a molecular weight of 172.18 g/mol, it is supplied at high purity for research use.
  • Potent KRasG12C inhibition demonstrated in cell assays.
  • Shows antiproliferative activity and anticancer effects in preclinical models.
  • Suitable as a research tool for KRas-driven signaling and lung cancer studies.
  • High purity (99.92%), minimizing impurities in experimental work.
  • Well-characterized physical properties (C11H8O2, 172.18 g/mol).
  • Available in gram-scale quantities for synthesis and biological testing.