Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-1-naphthaldehyde, MP Biomedicals™

Catalog No. ICN15129825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15129825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15129825 Supplier MP Biomedicals, Inc Supplier No. 0215129825

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 708-06-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
InChI Key NTCCNERMXRIPTR-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
PubChem CID 12819
IUPAC Name 2-hydroxynaphthalene-1-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2C=O)O

Specifications

Color Brown-Yellow
Quantity 25 g
Formula Weight 172.19
Physical Form Powder or Lumps
Chemical Name or Material 2-Hydroxy-1-naphthaldehyde
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.