Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-1-naphthaldehyde, tech.

Catalog No. AC121601000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC121601000 100 g
AC121605000 500 g
AC121600050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121601000 Supplier Thermo Scientific Chemicals Supplier No. 121601000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 708-06-5 | C11H8O2 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 708-06-5
MDL Number MFCD00004005
InChI Key NTCCNERMXRIPTR-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
PubChem CID 12819
IUPAC Name 2-hydroxynaphthalene-1-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2C=O)O

Specifications

Melting Point >77.0°C
Color Yellow
Boiling Point 192.0°C (27.0 mmHg)
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C11H8O2
Linear Formula HOC10H6CHO
Quantity 100 g
Beilstein 08,143
Solubility Information Solubility in water: 40mg/l
Molecular Weight (g/mol) 172.18
Formula Weight 172.18
Percent Purity ≥93% (on dry substance)
Grade Technical
Physical Form Moist Crystalline Powder
Chemical Name or Material 2-Hydroxy-1-naphthaldehyde, tech.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-902-0
RTECSNumber QJ0300000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.