Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-2-(trifluoromethyl)propionic acid, 94%

Catalog No. AAL1739903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1739903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1739903 Supplier Thermo Scientific Chemicals Supplier No. L1739903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 374-35-6 | C4H5F3O3 | 158.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-35-6
Molecular Formula C4H5F3O3
Molecular Weight (g/mol) 158.08
MDL Number MFCD00190646,MFCD03095400,MFCD07375449
InChI Key CTGJACFEVDCYMC-UHFFFAOYNA-N
Synonym 2-trifluoromethyl-2-hydroxypropionic acid, 3,3,3-trifluoro-2-hydroxy-2-methylpropionic acid, 2-hydroxy-2-trifluoromethyl propionic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl-propionic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl, methyl3,3,3-trifluoro-dl-lactate, acmc-1ai3k, acmc-209gc9, 2s-2-hydroxy-2-methyl-3,3,3-trifluoropropanoic acid, alpha-trifluoromethyl lactic acid
PubChem CID 2775125
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
SMILES CC(O)(C(O)=O)C(F)(F)F

Specifications

Melting Point ∼85°C
Boiling Point 189°C
Quantity 1 g
UN Number UN3261
Beilstein 1723691
Formula Weight 158.08
Percent Purity 94%
Chemical Name or Material 2-Hydroxy-2-(trifluoromethyl)propionic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 609-428-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.