Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-3-methoxybenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. H12735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12735G 5 g
H12731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12735G Supplier TCI America Supplier No. H12735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4383-05-5
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008362
InChI Key OSZHSESNQIMXMZ-UHFFFAOYSA-N
Synonym 2-(Hydroxymethyl)-6-methoxyphenol, 2-Hydroxy-3-(hydroxymethyl)anisole
PubChem CID 78088
IUPAC Name 2-(hydroxymethyl)-6-methoxyphenol
SMILES COC1=CC=CC(=C1O)CO

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3-methoxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.