Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate

Catalog No. AAH27604MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27604MD 250 mg
AAH2760403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27604MD Supplier Thermo Scientific Chemicals Supplier No. H27604MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 896437-63-1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 896437-63-1
MDL Number MFCD00078139
Synonym Quaternary ammonium beta-cylodextrin; QA-beta-CD

Specifications

Melting Point ∽280°C (decomposition)
Quantity 250 mg
Chemical Name or Material (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate
RTECSNumber GU2293000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.