Learn More
Medchemexpress LLC 2-hydroxy-3-nitro-5-(phenylmethyl)-benzaldehyde | 328565-16-8 | 99.9% | 257.24 g·mol-1 | C14H11NO4 | 25MG
SDP
Catalog No. 5000237760
Encompass_Preferred
Col003 is a small-molecule inhibitor of heat-shock protein 47 (Hsp47) that competitively binds the collagen-binding site and inhibits Hsp47 with an IC50 of 1.8 μM. It is supplied as a high-purity solid suitable for biochemical and cell-based studies investigating collagen-Hsp47 interactions, target validation, and assay development.
  • Selective inhibitor of Hsp47.
  • Reported IC50 of 1.8 μM.
  • Competitively binds the collagen-binding site on Hsp47.
  • High purity (99.87%) and characterized by analytical data.
  • Available in multiple small-mass pack sizes for research use.
  • Molecular weight 257.24 g·mol-1 and formula C14H11NO4.
Catalog No. 50-002-37760 Supplier Medchemexpress LLC Supplier No. HY12481725MG
Col003 is a small-molecule inhibitor of heat-shock protein 47 (Hsp47) that competitively binds the collagen-binding site and inhibits Hsp47 with an IC50 of 1.8 μM. It is supplied as a high-purity solid suitable for biochemical and cell-based studies investigating collagen-Hsp47 interactions, target validation, and assay development.
  • Selective inhibitor of Hsp47.
  • Reported IC50 of 1.8 μM.
  • Competitively binds the collagen-binding site on Hsp47.
  • High purity (99.87%) and characterized by analytical data.
  • Available in multiple small-mass pack sizes for research use.
  • Molecular weight 257.24 g·mol-1 and formula C14H11NO4.