Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxy-4-[4-[(5Z)-5-[(E)-2-methyl-3-phenylprop-2-enylidene]-4-oxo-2-sulfanylidene-1,3-thazolidin | 1164500-72-4 | MFCD03705956 | 96.1% | 482.57 | C24H22N2O5S2 | 1 ML
SDP

Catalog No. 5000240367
Encompass_Preferred

ML 145 is a small-molecule GPR35 antagonist for receptor pharmacology and cellular assays. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid quantities for experimental use.

  • GPR35 antagonist suitable for receptor pharmacology and cellular assays.
  • Reported purity 96.1% (HPLC).
  • CAS number 1164500-72-4.
  • Chemical formula C24H22N2O5S2, molecular weight 482.57 g·mol-1.
  • Solution format: 10 mM in DMSO, 1 mL; solids available in 5-100 mg sizes.
  • Recommended storage: powder at -20 °C; in solvent at -80 °C for long-term storage.
  • Provided with registry identifiers and structural descriptors for database cross-referencing.

Catalog No. 50-002-40367 Supplier Medchemexpress LLC Supplier No. HY10753610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML 145 is a small-molecule GPR35 antagonist for receptor pharmacology and cellular assays. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid quantities for experimental use.

  • GPR35 antagonist suitable for receptor pharmacology and cellular assays.
  • Reported purity 96.1% (HPLC).
  • CAS number 1164500-72-4.
  • Chemical formula C24H22N2O5S2, molecular weight 482.57 g·mol-1.
  • Solution format: 10 mM in DMSO, 1 mL; solids available in 5-100 mg sizes.
  • Recommended storage: powder at -20 °C; in solvent at -80 °C for long-term storage.
  • Provided with registry identifiers and structural descriptors for database cross-referencing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.